Learn More
Novus Biologicals™ LNX2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30925-25ug
 View more versions of this product
Product Code. 30067232
  • £162.00 / 25µg
Description
LNX2 Polyclonal antibody specifically detects LNX2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LNX2
Polyclonal
Western Blot : 1-5 μg/mL
FLJ23932, FLJ38000, ligand of Numb protein X 2, ligand of numb-protein X 2, MGC46315, Numb-binding protein 2, PDZ domain containing ring finger 1, PDZ domain-containing RING finger protein 1, PDZRN1FLJ12933
Synthetic peptide derived from human LNX2 protein
25 μg
Growth and Development, Neuronal Cell Markers, Zinc Finger
222484
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
Primary
Human
Purified