missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LNX2 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98102UV
This item is not returnable.
View return policy
Description
LNX2 Polyclonal antibody specifically detects LNX2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| LNX2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| FLJ23932, FLJ38000, ligand of Numb protein X 2, ligand of numb-protein X 2, MGC46315, Numb-binding protein 2, PDZ domain containing ring finger 1, PDZ domain-containing RING finger protein 1, PDZRN1FLJ12933 | |
| Produced in rabbits immunized with E. coli-derived Human LNX2 fragment. | |
| 0.1 mL | |
| Growth and Development, Neuronal Cell Markers, Zinc Finger | |
| 222484 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction