missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lnx1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-58844
This item is not returnable.
View return policy
Description
Lnx1 Polyclonal specifically detects Lnx1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| Lnx1 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| EC 6.3.2.-, ligand of numb-protein X 1ligand of numb-protein X, LNXmulti-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX, MPDZ, Numb-binding protein 1, PDZ domain-containing ring finger protein 2, PDZRN2E3 ubiquitin-protein ligase LNX | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| LNX1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:RSNRTRARPFERSTIRSRSFKKINRALSVLRRTKSGSAVANHADQGRENSENTTAPEVFPRLYHLIPDGEITSIKINRV | |
| 100 μL | |
| Cancer, Hypoxia, Lipid and Metabolism, Zinc Finger | |
| 84708 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction