missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LNPEP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534458
This item is not returnable.
View return policy
Description
A suggested positive control is human lung tissue lysate. PA5-34458 can be used with blocking peptide PEP-1500.
This gene encodes a zinc-dependent aminopeptidase that cleaves vasopressin, oxytocin, lys-bradykinin, met-enkephalin, dynorphin A and other peptide hormones. The protein can be secreted in maternal serum, reside in intracellular vesicles with the insulin-responsive glucose transporter GLUT4, or form a type II integral membrane glycoprotein. The protein catalyzes the final step in the conversion of angiotensinogen to angiotensin IV (AT4) and is also a receptor for AT4. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| LNPEP | |
| Polyclonal | |
| Unconjugated | |
| LNPEP | |
| 2010309L07Rik; 4732490P18Rik; aminopeptidase Vp165; angiotensin IV receptor; AT (4) receptor; CAP; Cystinyl aminopeptidase; GP160; insulin-regulated aminopeptidase; insulin-regulated membrane aminopeptidase; insulin-regulated membrane aminopeptidase IRAP; insulin-responsive aminopeptidase; IRAP; leucyl and cystinyl aminopeptidase; leucyl/cystinyl aminopeptidase; leucyl-cystinyl aminopeptidase; Leucyl-cystinyl aminopeptidase, pregnancy serum form; LNPEP; OTase; oxytocinase; Placental leucine aminopeptidase; PLAP; P-LAP; vasopressinase; vesicle protein of 165 kDa; vp160; vp165 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4012 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q9UIQ6 | |
| LNPEP | |
| A 15 amino acid synthetic peptide near the carboxy terminus of human LNPEP. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction