missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LNK/SH2B3 Antibody (OTI2D8), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72514PE
This item is not returnable.
View return policy
Description
LNK/SH2B3 Monoclonal specifically detects LNK/SH2B3 in Human samples. It is validated for Flow Cytometry.
Specifications
| LNK/SH2B3 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| SH2B3 | |
| Full length recombinant protein of human LNK/SH2B3 (NP_005466) produced in HEK293T cells. | |
| 0.1 mL | |
| 10019.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2D8 | |
| Flow Cytometry | |
| IDDM20, LNKSH2B adapter protein 3, Lymphocyte adapter protein, Lymphocyte-specific adapter protein Lnk, SH2B adaptor protein 3, Signal transduction protein Lnk | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction