missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LNK Polyclonal Antibody
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes a member of the SH2B adaptor family of proteins, which are involved in a range of signaling activities by growth factor and cytokine receptors. The encoded protein is a key negative regulator of cytokine signaling and plays a critical role in hematopoiesis. Mutations in this gene have been associated with susceptibility to celiac disease type 13 and susceptibility to insulin-dependent diabetes mellitus.
Specifications
Specifications
| Antigen | LNK |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | SH2B3 |
| Gene Accession No. | Q9UQQ2 |
| Gene Alias | AI429800; IDDM20; linker of T-cell receptor pathways; LNK; Lymphocyte adapter protein; lymphocyte-specific adapter protein Lnk; SH2B adapter protein 3; SH2B adaptor protein 3; Sh2b3; Signal transduction protein Lnk |
| Gene Symbols | SH2B3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?