missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMTK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585019
This item is not returnable.
View return policy
Description
Immunogen sequence: CSCLPLERGD AVAGWGGHPA LGCPHPPEDD SSLRAERGSL ADLPMAPPAS APPEFLDP Highest antigen sequence indentity to the following orthologs: Rat - 34%, Mouse - 91%.
Protein kinase which phosphorylates ESR1 (in vitro) and protects it against proteasomal degradation. May also regulate ESR1 levels indirectly via a PKC-AKT-FOXO3 pathway where it decreases the activity of PKC and the phosphorylation of AKT, thereby increasing binding of transcriptional activator FOXO3 to the ESR1 promoter and increasing ESR1 transcription (PubMed:21602804). Involved in endocytic trafficking of N-methyl-D-aspartate receptors (NMDAR) in neurons. [UniProt].
Specifications
| LMTK3 | |
| Polyclonal | |
| Unconjugated | |
| LMTK3 | |
| Aatyk3; Apoptosis-associated tyrosine kinase 3; BC059845; KIAA1883; Lemur tyrosine kinase 3; LMR3; Lmtk3; LOC691056; PPP1R101; protein phosphatase 1, regulatory subunit 101; serine/threonine-protein kinase LMTK3; similar to 5E5 antigen; TYKLM3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114783, 381983 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5XJV6, Q96Q04 | |
| LMTK3 | |
| Recombinant protein corresponding to Human LMTK3. Recombinant protein control fragment (Product #RP-108287). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction