missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMTK3 Antibody (6C5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00114783-M03
This item is not returnable.
View return policy
Description
LMTK3 Monoclonal antibody specifically detects LMTK3 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| LMTK3 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| AATYK3, EC 2.7.11.1, lemur tyrosine kinase 3KIAA1883TYKLM3LMR3, serine/threonine-protein kinase LMTK3 | |
| LMTK3 (XP_055866, 1151 a.a. ∽ 1250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VLVNGGLTPPKSEDKVSENGGLRFPRNTERPPETGPWRAPGPWEKTPESWGPAPTIGEPAPETSLERAPAPSAVVSSRNGGETAPGPLGPAPKNGTLEPG | |
| 0.1 mg | |
| Protein Kinase | |
| 114783 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 6C5 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| XP_055866 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction