missing translation for 'onlineSavingsMsg'
Learn More

LMTK3 Antibody (5A10), Novus Biologicals™

Product Code. 18403399 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18403399 0.05 mg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18403399 Supplier Novus Biologicals Supplier No. H00114783M06

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LMTK3 Monoclonal antibody specifically detects LMTK3 in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMTK3
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 5A10
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. XP_055866
Gene Alias AATYK3, EC 2.7.11.1, lemur tyrosine kinase 3KIAA1883TYKLM3LMR3, serine/threonine-protein kinase LMTK3
Host Species Mouse
Immunogen LMTK3 (XP_055866, 1151 a.a. ∽ 1250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. VLVNGGLTPPKSEDKVSENGGLRFPRNTERPPETGPWRAPGPWEKTPESWGPAPTIGEPAPETSLERAPAPSAVVSSRNGGETAPGPLGPAPKNGTLEPG
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 114783
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.