missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LMTK2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5111142

Product Code. 17129063

  • £550.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MLTGDTLSTS LQSSPEVQVP PTSFETEETP RRVPPDSLPT QGETQPTCLD VIVPEDCLHQ DISPDAVTVP VEILSTDART HSLDNRSQDS PGESEETLRL TESDSVLADD ILASRVSVGS SLPELGQELH NKPFSEDHH Highest antigen sequence indentity to the following orthologs: Mouse - 40%, Rat - 36%.

Protein kinases are enzymes that transfer a phosphate group from a phosphate donor, generally the g phosphate of ATP, onto an acceptor amino acid in a substrate protein. By this basic mechanism, protein kinases mediate most of the signal transduction in eukaryotic cells, regulating cellular metabolism, transcription, cell cycle progression, cytoskeletal rearrangement and cell movement, apoptosis, and differentiation. With more than 500 gene products, the protein kinase family is one of the largest families of proteins in eukaryotes. The family has been classified in 8 major groups based on sequence comparison of their tyrosine (PTK) or serine/threonine (STK) kinase catalytic domains. The tyrosine kinase (TK) group is mainly involved in the regulation of cell-cell interactions such as differentiation, adhesion, motility and death. There are currently about 90 TK genes sequenced, 58 are of receptor protein TK (e.g. EGFR, EPH, FGFR, PDGFR, TRK, and VEGFR families), and 32 of cytosolic TK (e.g. ABL, FAK, JAK, and SRC families).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LMTK2
Polyclonal
Unconjugated
LMTK2
2900041G10Rik; A330101P12Rik; aatyk2; Apoptosis-associated tyrosine kinase 2; brain-enriched kinase; BREK; CDK5/p35-regulated kinase; cprk; cyclin-dependent kinase 5/p35-regulated kinase; FLJ46659; hBREK; Kiaa1079; kinase phosphatase inhibitor 2; kinase/phosphatase/inhibitor 2; KPI2; KPI-2; KPI-2 protein; Lemur tyrosine kinase 2; LMKT2; LMR2; Lmtk2; PPP1R100; protein phosphatase 1, regulatory subunit 100; serine/threonine-protein kinase KPI-2; Serine/threonine-protein kinase LMTK2
Rabbit
Antigen affinity chromatography
RUO
22853
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.6 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8IWU2
LMTK2
Recombinant Protein corresonding to Human LMTK2. Recombinant protein control fragment (Product #RP-106019).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.