missing translation for 'onlineSavingsMsg'
Learn More

LMP7/PSMB8 Antibody [PE], Novus Biologicals™

Product Code. 30736241 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736241 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736241 Supplier Novus Biologicals Supplier No. AF7710P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

LMP7/PSMB8 Polyclonal antibody specifically detects LMP7/PSMB8 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMP7/PSMB8
Applications Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias beta5i, D6S216E, EC 3.4.25.1, LMP7MGC1491, Low molecular mass protein 7, low molecular weight protein 7, Macropain subunit C13, Multicatalytic endopeptidase complex subunit C13, protease component C13, proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctionalpeptidase 7), proteasome catalytic subunit 3i, Proteasome component C13, proteasome subunit beta 5i, proteasome subunit beta type-8, Proteasome subunit beta-5i, proteasome subunit Y2, proteasome-related gene 7, PSMB5iD6S216, Really interesting new gene 10 protein, RING10proteasome (prosome, macropain) subunit, beta type, 8 (large multifunctionalprotease 7), Y2
Host Species Sheep
Immunogen E. coli-derived recombinant human LMP7/PSMB8, Thr73-Gln276, Accession # P28062
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 5696
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.