missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMOD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546523
This item is not returnable.
View return policy
Description
Peptide sequence: VKSEEKTQEE HEEIEKRNKN MAQYLKEKLN NEIVANKRES KGSSNIQETD Sequence homology: Cow: 77%; Dog: 77%; Horse: 79%; Human: 100%.
The function of this protein remains unknown.
Specifications
| LMOD3 | |
| Polyclonal | |
| Unconjugated | |
| LMOD3 | |
| 5430424A14Rik; leiomodin 3; leiomodin 3 (fetal); leiomodin, fetal form; Leiomodin-3; LMOD3; NEM10; RGD1564924 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 56203 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q0VAK6 | |
| LMOD3 | |
| Synthetic peptide directed towards the N-terminal of human LMOD3 (aa 95-144). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction