missing translation for 'onlineSavingsMsg'
Learn More

LMOD1 Antibody (3B8), Novus Biologicals™

Product Code. 18381359 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18381359 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18381359 Supplier Novus Biologicals Supplier No. H00025802M08

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LMOD1 Monoclonal antibody specifically detects LMOD1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMOD1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3B8
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH01755
Gene Alias 64 kDa autoantigen 1D, 64 kDa autoantigen 1D3,1D, 64 kDa autoantigen D1,64kD, D1, FLJ55689, leimodin 1 (smooth muscle), leiomodin 1 (smooth muscle), Leiomodin, muscle form, leiomodin-1, SM-Lmod, Smooth muscle leiomodin, thyroid and eye muscle autoantigen D1 (64kD), Thyroid-associated ophthalmopathy autoantigen
Host Species Mouse
Immunogen LMOD1 (AAH01755, 1 a.a. ~ 269 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEELEKELDVVDPDGSVPVGLRQRNQTEKQSTGVYNREAMLNFCEKETKKLMQREMSMDESKQVETKTDAKNGEERGRDASKKALGPRRDSDLGKEPKRGGLKKSFSRDRDEAGGKSGEKPKEEKIIRGIDKGRVRAAVDKKEAGKDGRGEERAVATKKEEEKKGSDRNTGLSRDKDKKREEMKEVAKKEDDEKVKGGAPAAPPPPPPPLAPPLIMENLKNSLSPATQRKMGDKVLPAQEKNSRDQLLAAIRSSNLKQLKKVEVPKLLQ
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 25802
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.