missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMO4 Rabbit anti-Human, Mouse, Rat, Clone: 5C0I1, Novus Biologicals™
Rabbit Monoclonal Antibody
£158.00 - £395.00
Specifications
| Antigen | LMO4 |
|---|---|
| Clone | 5C0I1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18350616
|
Novus Biologicals
NBP3-16630-20UL |
20 μg |
£158.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18343316
|
Novus Biologicals
NBP3-16630-100UL |
100 μg |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LMO4 Monoclonal antibody specifically detects LMO4 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| LMO4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Tumor Suppressors | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 8543 | |
| IgG | |
| Affinity purified |
| 5C0I1 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Breast tumor autoantigen, LIM domain only 4, LIM domain only protein 4, LIM domain transcription factor LMO4, LMO-4 | |
| A synthetic peptide corresponding to a sequence within amino acids 66-165 of human LMO4 (P61968). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title