missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMO4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £513.00
Specifications
| Antigen | LMO4 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619717
|
Novus Biologicals
NBP2-49342-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692749
|
Novus Biologicals
NBP2-49342 |
0.1 mL |
£513.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LMO4 Polyclonal antibody specifically detects LMO4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| LMO4 | |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Tumor Suppressors | |
| PBS (pH 7.2), 40% Glycerol | |
| 8543 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Breast tumor autoantigen, LIM domain only 4, LIM domain only protein 4, LIM domain transcription factor LMO4, LMO-4 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FTCSTCRNRLVPGDRFHYINGSLFCEHDRPTALINGHLNSLQSNPLLPDQKVC | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title