missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LMO4 Monoclonal antibody specifically detects LMO4 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | LMO4 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 3B4 |
| Conjugate | Unconjugated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_006760 |
| Gene Alias | Breast tumor autoantigen, LIM domain only 4, LIM domain only protein 4, LIM domain transcription factor LMO4, LMO-4 |
| Host Species | Mouse |
| Immunogen | LMO4 (NP_006760, 57 a.a. ~ 165 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LGDIGTSCYTKSGMILCRNDYIRLFGNSGACSACGQSIPASELVMRAQGNVYHLKCFTCSTCRNRLVPGDRFHYINGSLFCEHDRPTALINGHLNSLQSNPLLPDQKVC |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?