missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMO2 Rabbit anti-Human, Clone: LMO2/3147R, Novus Biologicals™
Rabbit Monoclonal Antibody
£245.00
Specifications
| Antigen | LMO2 |
|---|---|
| Clone | LMO2/3147R |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml |
| Applications | Flow Cytometry, Immunofluorescence |
Description
LMO2 Monoclonal specifically detects LMO2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
| LMO2 | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 4005 | |
| IgG | |
| Protein A or G purified |
| LMO2/3147R | |
| Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2 | |
| Recombinant fragment (around aa 23-140) human LMO2 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title