Learn More
Invitrogen™ LMO2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116908
Description
Positive Samples: LO2, A-549, 293T, Mouse liver, Mouse kidney, Rat liver Immunogen sequence: MSSAIERKSL DPSEEPVDEV LQIPPSLLTC GGCQQNIGDR YFLKAIDQYW HEDCLSCDLC GCRLGEVGRR LYYKLGRKLC RRDYLRLFGQ DGLCASCDKR IRAYEMTMRV KDKVYHLECF KCAACQKHFC VGDRYLLINS DIVCEQDIYE WTKINGMI.
LMO2 encodes a cysteine-rich, two LIM-domain protein that is required for yolk sac erythropoiesis. The LMO2 protein has a central and crucial role in hematopoietic development and is highly conserved. The LMO2 transcription start site is located approximately 25 kb downstream from the 11p13 T-cell translocation cluster (11p13 ttc), where a number T-cell acute lymphoblastic leukemia-specific translocations occur. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| LMO2 | |
| Polyclonal | |
| Unconjugated | |
| Lmo2 | |
| cysteine-rich protein TTG-2; LIM domain only 2; LIM domain only 2 (rhombotin-like 1); LIM domain only protein 2; LIM domain only-2; LIM only 2; LMO2; LMO-2; LMO2 protein; Rbtn2; Rbtn-2; RBTNL1; RHOM2; Rhom-2; rhombotin 2; rhombotin-2; rhombotin-like 1; si:dkey-10o6.5; T-cell translocation gene 2; T-cell translocation protein 2; TTG2; wu:fc83a08; zgc:111930; zlmo2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16909, 362176, 4005 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P25791, P25801 | |
| Lmo2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-158 of human LMO2 (NP_0011357881). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.