missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMO2 Mouse anti-Human, Clone: LMO2/1971, Novus Biologicals™
Mouse Monoclonal Antibody
£377.00
Specifications
| Antigen | LMO2 |
|---|---|
| Clone | LMO2/1971 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) |
Description
LMO2 Monoclonal specifically detects LMO2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin.Specifications
| LMO2 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer | |
| 10 mM PBS with 0.05% BSA | |
| 4005 | |
| IgG2b κ | |
| Protein A or G purified |
| LMO2/1971 | |
| Western Blot 1 to 2 μg/mL, Flow Cytometry 1 to 2 μg/million cells, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2 | |
| Recombinant human LMO2 protein fragment (around aa 23-140) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title