missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMO2 Antibody (LMO2/1971), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08268CL1
This item is not returnable.
View return policy
Description
LMO2 Monoclonal antibody specifically detects LMO2 in Human samples. It is validated for Flow Cytometry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| LMO2 | |
| Monoclonal | |
| CoraFluor 1 | |
| Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2 | |
| Recombinant human LMO2 protein fragment (around aa 23-140) (exact sequence is proprietary) (Uniprot: P25791) | |
| 0.1 mL | |
| Angiogenesis, Cancer | |
| 4005 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin) | |
| LMO2/1971 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction