missing translation for 'onlineSavingsMsg'
Learn More

LMO2 Antibody (9H9), Novus Biologicals™

Rabbit Monoclonal Antibody

£260.00 - £406.00

Specifications

Antigen LMO2
Clone 9H9
Dilution Western Blot 1:500-1:2000, ELISA
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30034956
View Documents
Novus Biologicals
NBP3-26166-50ul
50 μL
£260.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30034454
View Documents
Novus Biologicals
NBP3-26166-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LMO2 Monoclonal antibody specifically detects LMO2 in Human samples. It is validated for Western Blot, ELISA
Specifications

Specifications

LMO2
Western Blot 1:500-1:2000, ELISA
Monoclonal
Purified
RUO
Human
Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2
A synthesized peptide derived from Human LMO2 [UniProt P25791]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
9H9
Western Blot, ELISA
Unconjugated
Rabbit
Angiogenesis, Cancer
PBS, pH 7.4, 150mM NaCl and 50% glycerol
4005
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.