missing translation for 'onlineSavingsMsg'
Learn More

LMO2 Antibody (1A9-3B11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication

£183.00 - £415.00

Specifications

Antigen LMO2
Clone 1A9-3B11
Concentration 1.0 mg/mL
Dilution Western Blot 0.5 ug/ml, Flow Cytometry 1-2 ug/ml per million cells, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500, Immunofluorescence, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18254160
View Documents
Novus Biologicals
NB110-78626SS
0.025 mL
£183.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18757563
View Documents
Novus Biologicals
NB110-78626
0.1 mL
£415.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LMO2 Monoclonal specifically detects LMO2 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications

Specifications

LMO2
1.0 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Mouse
Angiogenesis, Cancer
P25791
4005
Recombinant human LMO2. [UniProt# P25791]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1A9-3B11
Western Blot 0.5 ug/ml, Flow Cytometry 1-2 ug/ml per million cells, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500, Immunofluorescence, Chromatin Immunoprecipitation (ChIP) 1:10-1:500
Monoclonal
Purified
RUO
Human
Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2
LMO2
IgG1 κ
Protein G purified
24 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.