missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMK-235, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
Description
LMK-235 is a potent and selective HDAC4/5 inhibitor, inhibits HDAC5, HDAC4, HDAC6, HDAC1, HDAC2, HDAC11 and HDAC8, with IC50s of 4.22 nM, 11.9 nM, 55.7 nM, 320 nM, 881 nM, 852 nM and 1278 nM, respectively, and is used in cancer research.
Specifications
Specifications
| Chemical Name or Material | LMK-235 |
| Physical Form | Solid |
| CAS | 1418033-25-6 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C15H22N2O4 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : ≥ 30 mg/mL (101.92 mM) |
| SMILES | O=C(NOCCCCCC(NO)=O)C1=CC(C)=CC(C)=C1 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction