missing translation for 'onlineSavingsMsg'
Learn More

LMK-235, MedChemExpress

Product Code. 16483687
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483687

Brand: Medchem Express HY18998

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LMK-235 is a potent and selective HDAC4/5 inhibitor, inhibits HDAC5, HDAC4, HDAC6, HDAC1, HDAC2, HDAC11 and HDAC8, with IC50s of 4.22 nM, 11.9 nM, 55.7 nM, 320 nM, 881 nM, 852 nM and 1278 nM, respectively, and is used in cancer research.

LMK-235 is a potent and selective HDAC4/5 inhibitor, inhibits HDAC5, HDAC4, HDAC6, HDAC1, HDAC2, HDAC11 and HDAC8, with IC50s of 4.22 nM, 11.9 nM, 55.7 nM, 320 nM, 881 nM, 852 nM and 1278 nM, respectively, and is used in cancer research.

Specifications

Chemical Name or Material LMK-235
Physical Form Solid
CAS 1418033-25-6
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C15H22N2O4
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 30 mg/mL (101.92 mM)
SMILES O=C(NOCCCCCC(NO)=O)C1=CC(C)=CC(C)=C1
Molecular Weight (g/mol) 294.35
Formula Weight 294.35
Percent Purity 98.1%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.