missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMF2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16249-1-AP-150UL
This item is not returnable.
View return policy
Description
Involved in the maturation of specific proteins in the endoplasmic reticulum. May be required for maturation and transport of active lipoprotein lipase (LPL) through the secretory pathway.Specifications
| LMF2 | |
| Polyclonal | |
| Unconjugated | |
| LMF2 | |
| lipase maturation factor 2, LMF2, TMEM112B, TMEM153, Transmembrane protein 112B, Transmembrane protein 153 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 105847, 315218, 91289 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A1L1J9, Q8C3X8, Q9BU23 | |
| Lmf2 | |
| LMF2 Fusion Protein Ag9208 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction