missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The protein encoded by this gene resides in the endoplasmic reticulum, and is involved in the maturation and transport of lipoprotein lipase through the secretory pathway. Mutations in this gene are associated with combined lipase deficiency.
Specifications
Specifications
| Antigen | LMF1 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | LMF1 |
| Gene Accession No. | Q3U3R4, Q96S06 |
| Gene Alias | C16orf26, HMFN1876, JFP11, lipase maturation factor 1, LMF1, TMEM112, TMEM112A, Transmembrane protein 112 |
| Gene Symbols | Lmf1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?