missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMBRD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590773
This item is not returnable.
View return policy
Description
Immunogen sequence: PLNLIKGTRS AAYERLENTE DIEEVEQHIQ TIKSKSKDGR PLPARDKRAL KQFEERLRTL KKRERHLEFI ENSWWTKFCG A; Positive Samples: U-87MG, HeLa, Mouse brain, Mouse lung, Mouse spleen; Cellular Location: Lysosome membrane, Multi-pass membrane protein.
LMBRD1, also known as NESI (nuclear export signal-interacting protein, is a lysosomal membrane protein that is thought be involved in the transport and metabolism of cobalamin. LMBRD1 was initially identified as interacting with the large form of the hepatitis delta antigen and may be required for the nucleocytoplasmic shuttling of the hepatitis delta virus. Mutations in this gene are associated with the vitamin B12 metabolism disorder termed, homocystinuria-megaloblastic anemia complementation type F (cblF).
Specifications
| LMBRD1 | |
| Polyclonal | |
| Unconjugated | |
| Lmbrd1 | |
| 0910001K20Rik; AV347960; BM-021; C6orf209; CD001; HDAg-L-interacting protein NESI; hepatitis delta antigen-L interacting protein; liver regeneration p-53 related protein; LMBD1; LMBR1 domain containing 1; LMBR1 domain-containing protein 1; LMBRD1; MAHCF; MSTP044; NESI; nuclear export signal-interacting protein; Probable lysosomal cobalamin transporter; protein N90b | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 55788, 68421 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.39 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8K0B2, Q9NUN5 | |
| Lmbrd1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 220-300 of human LMBRD1 (NP_0608383). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction