missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMBRD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£196.00 - £470.00
Specifications
| Antigen | LMBRD1 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18485461
|
Novus Biologicals
NBP1-88729-25ul |
25 μL |
£196.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18215168
|
Novus Biologicals
NBP1-88729 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LMBRD1 Polyclonal specifically detects LMBRD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| LMBRD1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 55788 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:KGTRSAAYERLENTEDIEEVEQHIQTIKSKSKDGRPLPARDKRALKQFEERLRTLKKRERHLEFIENSWWTKFCGALRP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| bA810I22.1, C6orf209, chromosome 6 open reading frame 209, FLJ11240, FLJ11854, HDAg-L-interacting protein NESI, hepatitis delta antigen-L interacting protein, liver regeneration p-53 related protein, LMBD1, LMBR1 domain containing 1, LMBR1 domain-containing protein 1, NESIcblF, Nuclear export signal-interacting protein, probable lysosomal cobalamin transporter | |
| LMBRD1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title