missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMBR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543411
This item is not returnable.
View return policy
Description
Peptide sequence: MEGQDEVSAR EQHFHSQVRE STICFLLFAI LYVVSYFIIT RYKRKSDEQE Sequence homology: Cow: 100%; Dog: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
This gene encodes a member of the LMBR1-like membrane protein family. Another member of this protein family has been shown to be a lipocalin transmembrane receptor. A highly conserved, cis-acting regulatory module for the sonic hedgehog gene is located within an intron of this gene. Consequently, disruption of this genic region can alter sonic hedgehog expression and affect limb patterning, but it is not known if this gene functions directly in limb development. Mutations and chromosomal deletions and rearrangements in this genic region are associated with acheiropody and preaxial polydactyly, which likely result from altered sonic hedgehog expression.
Specifications
| LMBR1 | |
| Polyclonal | |
| Unconjugated | |
| Lmbr1 | |
| 1110048D14Rik; ACHP; AU017641; C7orf2; DIF14; Differentiation-related gene 14 protein; FLJ11665; limb development membrane protein 1; limb region 1; limb region 1 homolog; limb region 1 protein; Limb region 1 protein homolog; Lmbr1; LSS; PPD2; THYP; TPT; Unknown (protein for MGC:134350); ZRS | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64327 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WVP7 | |
| Lmbr1 | |
| Synthetic peptide directed towards the N-terminal of human LMBR1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction