missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMAN2 Antibody (02), HRP , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05830H
This item is not returnable.
View return policy
Description
LMAN2 Monoclonal antibody specifically detects LMAN2 in Human samples. It is validated for Western Blot, ELISA, ELISA, ELISA
Specifications
| LMAN2 | |
| Monoclonal | |
| HRP | |
| C5orf8, chromosome 5 open reading frame 8, Glycoprotein GP36b, GP36B, Lectin mannose-binding 2, lectin, mannose-binding 2, vesicular integral protein of 36 kDa, vesicular integral-membrane protein VIP36, VIP36 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LMAN2 (Q12907; Asp 45-Arg 322). | |
| 0.1 mL | |
| Signal Transduction | |
| 10960 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| 02 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction