Learn More
LMAN2 Antibody (02), DyLight 680 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-05830FR
Product Code. 18465549
  • £329.00 / 0.10mL
Description
LMAN2 Monoclonal antibody specifically detects LMAN2 in Human samples. It is validated for Western Blot, ELISA, ELISA, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LMAN2
Monoclonal
DyLight 680
C5orf8, chromosome 5 open reading frame 8, Glycoprotein GP36b, GP36B, Lectin mannose-binding 2, lectin, mannose-binding 2, vesicular integral protein of 36 kDa, vesicular integral-membrane protein VIP36, VIP36
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human LMAN2 (Q12907; Asp 45-Arg 322).
0.1 mL
Signal Transduction
10960
Store at 4°C in the dark.
IgG
Western Blot, ELISA
02
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified