missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMAN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13364-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.Specifications
| LMAN1 | |
| Polyclonal | |
| Unconjugated | |
| LMAN1 | |
| ERGIC 53, ERGIC-53, F5F8D, FMFD1, gp58, Lectin mannose binding 1, lectin, mannose binding, 1, LMAN1, MCFD1, MR60, Protein ERGIC 53 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116666, 3998, 70361 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49257, Q62902, Q9D0F3 | |
| LMAN1 | |
| ERGIC-53 Fusion Protein Ag4183 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction