missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMAN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
£155.00 - £430.00
Specifications
| Antigen | LMAN1 |
|---|---|
| Concentration | 0.64 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17770654
![]() |
Invitrogen™
PA5119250 |
200 μL |
£430.00
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17723194
|
Invitrogen™
PA5121794 |
20 μL |
£155.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LMAN1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.Specifications
| LMAN1 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| LMAN1 | |
| 2610020P13Rik; AI326273; AU043785; C730041J05; EGK_09725; endoplasmic reticulum-golgi intermediate compartment protein 53; ERGIC53; ERGIC-53; ER-Golgi intermediate compartment 53 kDa protein; F5F8D; FMFD1; Gp58; intracellular mannose-specific lectin MR60; Lectin mannose-binding 1; lectin, mannose binding 1; lectin, mannose-binding, 1; LMAN1; MCFD1; MR60; P58; Protein ERGIC-53 | |
| LMAN1 | |
| Primary | |
| Antigen affinity chromatography, Protein A |
| 0.64 mg/mL | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P49257 | |
| 3998 | |
| Recombinant protein Lectin, Mannose Binding 1. The antigen corresponds to amino acid range 198-475 of the target protein. | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title