missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Invitrogen™ LMAN1 Monoclonal Antibody (OTI1C5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525373
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
LMAN1 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.
Tekniske data
| LMAN1 | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49257 | |
| LMAN1 | |
| Full length human recombinant protein of LMAN1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1C5 | |
| Unconjugated | |
| LMAN1 | |
| 2610020P13Rik; AI326273; AU043785; C730041J05; EGK_09725; endoplasmic reticulum-golgi intermediate compartment protein 53; ERGIC53; ERGIC-53; ER-Golgi intermediate compartment 53 kDa protein; F5F8D; FMFD1; Gp58; intracellular mannose-specific lectin MR60; Lectin mannose-binding 1; lectin, mannose binding 1; lectin, mannose-binding, 1; LMAN1; MCFD1; MR60; P58; Protein ERGIC-53 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3998 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion