missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LLPH Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | LLPH |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18362017
|
Novus Biologicals
NBP3-04428-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18394796
|
Novus Biologicals
NBP3-04428-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LLPH Polyclonal specifically detects LLPH in Human, Rat samples. It is validated for Western Blot.Specifications
| LLPH | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 84298 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| C12orf31, Chromosome 12 Open Reading Frame 31, CPERP-G, HLLP, Human LAPS18-Like Protein, LLP Homolog, Long-Term Synaptic Facilitation, LLP Homolog, Long-Term Synaptic Facilitation (Aplysia), Protein LAPS18-Like, Protein LLP Homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-129 of human LLPH (NP_115714.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title