missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
LLPH Polyclonal specifically detects LLPH in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | LLPH |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, pH7.3. |
| Gene Alias | C12orf31, Chromosome 12 Open Reading Frame 31, CPERP-G, HLLP, Human LAPS18-Like Protein, LLP Homolog, Long-Term Synaptic Facilitation, LLP Homolog, Long-Term Synaptic Facilitation (Aplysia), Protein LAPS18-Like, Protein LLP Homolog |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-129 of human LLPH (NP_115714.1). |
| Purification Method | Affinity purified |
| Quantity | 20 μL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?