missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LLPH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565532
This item is not returnable.
View return policy
Description
Immunogen sequence: KMQCEVKDEKD DMKMETDIKR NKKTLLDQHG QYPIWMNQRQ RKRLKAKREK RKGKSKAKAV KVAKG Highest antigen sequence identity to the following orthologs - mouse 72%, rat 81%.
In hippocampal neurons, regulates dendritic and spine growth and synaptic transmission.
Specifications
| LLPH | |
| Polyclonal | |
| Unconjugated | |
| LLPH | |
| C12orf31; cPERP-G; hLLP; human LAPS18-like protein; LLP homolog, long-term synaptic facilitation; LLP homolog, long-term synaptic facilitation (Aplysia); LLPH; Protein LAPS18-like; Protein LLP homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84298 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BRT6 | |
| LLPH | |
| Recombinant Human LLPH. Recombinant protein control fragment (Product #RP-106207). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction