missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LLPH Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109921
This item is not returnable.
View return policy
Description
Immunogen sequence: MAKSLRSKWK RKMRAEKRKK NAPKEASRLK SILKLDGDVL MKDVQEIATV VVPKPKHCQE KMQCEVKDEK DDMKMETDIK RNKKTLLDQH GQYPIWMNQR QRKRLKAKRE KRKGKSKAKA VKVAKGLAW.
In hippocampal neurons, regulates dendritic and spine growth and synaptic transmission.
Specifications
| LLPH | |
| Polyclonal | |
| Unconjugated | |
| LLPH | |
| C12orf31; cPERP-G; hLLP; human LAPS18-like protein; LLP homolog, long-term synaptic facilitation; LLP homolog, long-term synaptic facilitation (Aplysia); LLPH; Protein LAPS18-like; Protein LLP homolog | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 299818, 84298 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.57 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9BRT6 | |
| LLPH | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-129 of human LLPH (NP_115714.1). | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction