missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LLGL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109920
This item is not returnable.
View return policy
Description
Immunogen sequence: RQVFVKCTLH PSDQLALEGP LSRVKSLKKS LRQSFRRMRR SRVSSHKRRP GGPTGEAQAQ AVNIKAERTG LQNMELAPVQ RKIEARSAED SFTGFVRTLY FADTYLRDSS RHCPSLWAGT NGGTVYAFSL RVPPAERRTD EPVRAEQAKE IQLMHRAPVV GILVLDGHNV PLPEPLEVAH DLSKSPDMQG SHQLLVVSEE.
LLGL1 is a protein that is similar to a tumor suppressor in Drosophila. The protein is part of a cytoskeletal network and is associated with nonmuscle myosin II heavy chain and a kinase that specifically phosphorylates this protein at serine residues. The gene for LLGL1 is located within the Smith-Magenis syndrome region on chromosome 17. LLGL2 is a protein similar to lethal (2) giant larvae of Drosophila. In fly, the protein's ability to localize cell fate determinants is regulated by the atypical protein kinase C (aPKC). In human, this protein interacts with aPKC-containing complexes and is cortically localized in mitotic cells.
Specifications
| LLGL2 | |
| Polyclonal | |
| Unconjugated | |
| LLGL2 | |
| 9130006H11Rik; HGL; Hugl-2; human giant larvae homolog; lethal giant larvae homolog 2; lethal giant larvae homolog 2 (Drosophila); lethal giant larvae homolog 2, scribble cell polarity complex component; lethal giant larvae-like protein 2; Lethal(2) giant larvae protein homolog 2; LGL2; LLGL scribble cell polarity complex component 2; Llgl2; LLGL2, scribble cell polarity complex component; Llglh2; RGD1560307 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 217325, 360661, 3993 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3TJ91, Q6P1M3 | |
| LLGL2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 620-819 of mouse LLGL2 (NP_663413.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction