missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX LKB1/STRADα/MO25α Protein, Active

Product Code. p-7228301
Encompass_Preferred
Click to view available options
Quantity:
10 μg
125 μg
Unit Size:
10µg
125µg
This item is not returnable. View return policy

Product Code. 17320404

Brand: Eurofins DiscoverX 14596

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, recombinant full-length human LKB1 with an N-terminal 6His tag. Provided as a complex with GST-STRADα and GST-MO25α, for use in Kinase Assays.

LKB1 is a tumor suppressor serine/threonine-protein kinase that controls the activity of AMP-activated protein kinase (AMPK) family members, thereby playing a role in various processes such as cell metabolism, cell polarity, apoptosis and DNA damage response. Mutations in this gene have been associated with the autosomal dominant Peutz-Jeghers syndrome, as well as with skin, pancreatic, and testicular cancers.

GenBank NM000455 / GenBank AF308302 / GenBank NM_016289

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 52.1 kDa (LKB1), 76.1 kDa (STRADα) and 66.8 kDa (MO25α)
Quantity 10 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His / N-terminal GST / N-terminal GST
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Full-length
Purity or Quality Grade Combined ≥80%
Protein LKB1/STRADα/MO25α, active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.