Learn More
Eurofins DiscoverX LKB1/STRADα/MO25α Protein, Active
Description
LKB1 is a tumor suppressor serine/threonine-protein kinase that controls the activity of AMP-activated protein kinase (AMPK) family members, thereby playing a role in various processes such as cell metabolism, cell polarity, apoptosis and DNA damage response. Mutations in this gene have been associated with the autosomal dominant Peutz-Jeghers syndrome, as well as with skin, pancreatic, and testicular cancers.
GenBank NM000455 / GenBank AF308302 / GenBank NM_016289
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 52.1 kDa (LKB1), 76.1 kDa (STRADα) and 66.8 kDa (MO25α) |
| Quantity | 125 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His / N-terminal GST / N-terminal GST |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | Combined ≥80% |
| Protein | LKB1/STRADα/MO25α, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.