missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LKAP Antibody, Novus Biologicals™

Product Code. 18348248 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18348248 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18348248 Supplier Novus Biologicals™ Supplier No. H00009665B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

LKAP Polyclonal antibody specifically detects LKAP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LKAP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. ENSP00000300014
Gene Alias KIAA0430, limkain-b1, LKAP
Host Species Mouse
Immunogen KIAA0430 (ENSP00000300014, 1 a.a. - 581 a.a.) full-length human protein. MGSKRLLTLTHRAQVKRFTQDLLKLLKSQASKQVIVREFSQAYHWCFSKDWDVTEYGVCELIDIVSEIPDTTICLSQQDNEMVICIPKRERTQDEIERTKQFSKDVVDLLRHQPHFRMPFNKFIPSYHHHFGRQCKLAYYGFTKLLELFEAIPDTLQVLECGEEKILTLTEVERFKALAAQFVKLLRSQKDNCLMMTDLLTEYAKTFGYTFRLQDYDVSSISALTQKLCHVVKVADIESGRQIQLINRKSLRSLTAQLLVLLMSWEGTTHLSVEELKRHYESTHNTPLNPCEYGFMTLTELLKSLPYLVEVFTNDKMEECVKLTSLYLFAKNVRSLLHTYHYQQIFLHEFSMAYTKYVGETLQPKTYGHSSVEELLGAIPQVVWIKGHGHKRIVVLKNDMKSRLSSLSLSPANHENQPSEGERILEVPESHTASELKLGADGSGPSHTEQELLRLTDDSPVDLLCAPVPSCLPSPQLRPDPVILQSADLIQFEERPQEPSEIMILNQEEKMEIPIPGKSKTLTSDSSSSCISAAVPVPPCPSSETSESLLSKDPVESPAKKQPKNRVKLAANFSLAPITKL
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9665
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.