missing translation for 'onlineSavingsMsg'
Learn More

LKAP Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18411158 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18411158 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18411158 Supplier Novus Biologicals Supplier No. NBP222257JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LKAP Polyclonal antibody specifically detects LKAP in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen LKAP
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA0430, limkain-b1, LKAP
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1625 and 1675 of human Limkain b1 using the numbering given in entry NP_055462.2 (GeneID 9665).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9665
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.