missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIX1 Antibody (RM0282-4F47), DyLight 550, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-11779R
This item is not returnable.
View return policy
Description
LIX1 Monoclonal specifically detects LIX1 in Mouse samples. It is validated for Western Blot.
Specifications
| LIX1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LIX1 | |
| Mouse recombinant LIX1 | |
| 0.1 mL | |
| 167410 | |
| Store at 4C in the dark. | |
| IgG2 |
| Western Blot | |
| RM0282-4F47 | |
| Western Blot | |
| C5orf11, chromosome 5 open reading frame 11, FLJ25534, limb expression 1, Lix1 homolog (chicken), Lix1 homolog (mouse), protein limb expression 1 homolog | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction