missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat LIX (CXCL5) ELISA Kit

Product Code. p-7230666
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465373 96 Tests 1 set
17135919 5 x 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465373 Supplier Invitrogen™ Supplier No. ERCXCL5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

The Rat LIX (CXCL5) ELISA quantitates Rt LIX in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt LIX. Principle of the method The Rat LIX solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is an inflammatory chemokine that belongs to the CXC chemokine family. This chemokine is produced concomitantly with interleukin-8 (IL8) in response to stimulation with either interleukin-1 (IL1) or tumor necrosis factor-alpha (TNFA). This chemokine is a potent chemotaxin involved in neutrophil activation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P97885
Assay Range 24.69 to 6000 pg/mL
Assay Sensitivity 20 pg/mL
Calibration Range 20 pg/mL to 6000 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate HRP
Product Type ELISA
Protein Family Chemokine & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with any of the cytokines tested: rat CINC-2, CINC-3, CNTF, Fractalkine, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-10, GM-CSF, IFN-gamma, Leptin, MCP-1, MIP-3 alpha, beta-NGF, TIMP-1, TNF-alpha, VEGF.
Sufficient For 96 assays
Target Species Rat
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Colorimetric Microplate Reader
Gene Symbol Cxcl5, Cxcl6
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Immunology
Sample Volume Plasma 33 μL, Serum 33 μL, Supernatant 100 μL
Storage Requirements 2°C to 8°C
Synonym Cxcl5
Target CXCL5
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.