Learn More
Invitrogen™ Rat LIX-Cell Lysate (CXCL5) ELISA Kit
Beschreibung
The Rat LIX (CXCL5) ELISA quantitates Rt LIX in rat cell or tissue lysates. The assay will exclusively recognize both natural and recombinant Rt LIX. Principle of the method The Rat LIX solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Spezifikation
Spezifikation
| Accession Number | P97885 |
| Assay Range | 8.23 to 6000 pg/mL |
| Assay Sensitivity | 15 pg/ml |
| Calibration Range | 15 pg/mL to 6000 pg/mL |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Product Type | ELISA |
| Protein Family | Chemokine & Receptors |
| Sample Type | Cell Lysates |
| Specificity | The antibody pair provided in this kit recognizes rat LIX / CXCL5. |
| Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.