missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Livin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587263
This item is not returnable.
View return policy
Description
Purity is > 95% by SDS-PAGE.
The inhibitor of apoptosis (IAP) family of proteins regulates programmed cell death triggered by various stimuli. All IAPs have at least one baculovirus IAP repeat (BIR) motif that is essential for their anti-apoptotic activity. Recently, it has been identified that a novel human inhibitor of apoptosis protein (IAP) family member termed Livin or ML-IAP, which contains a single baculoviral IAP repeat (BIR) domain and a COOH-terminal RING finger domain. Livin gene has two splicing variants that contain open reading frames of 298 and 280 amino acids and both contained a single copy of baculovirus IAP repeat (BIR) and RING domain. Both of the isoforms inhibit TNFa induced apoptosis in Jurkat cells. Livin expression is low in adult tissues. It is relatively expressed at a higher level in developmental tissues and in many cancer cells.
Specifications
| Livin | |
| Polyclonal | |
| Unconjugated | |
| BIRC7 | |
| baculoviral IAP repeat containing 7; baculoviral IAP repeat-containing 7; baculoviral IAP repeat-containing 7 (livin); baculoviral IAP repeat-containing protein 7; Baculoviral IAP repeat-containing protein 7 30 kDa subunit; Baculoviral IAP repeat-containing protein 7 30kDa subunit; Birc7; E130019N06; KIAP; Kidney inhibitor of apoptosis protein; LIVIN; livin inhibitor of apoptosis; melanoma inhibitor of apoptosis protein; MLIAP; ML-IAP; RGD1562883; RING finger protein 50; RING-type E3 ubiquitin transferase BIRC7; RNF50; RP11-261N11.7; Truncated livin; UNQ5800/PRO19607/PRO21344 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 296468, 329581, 79444 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| A2AWP0, Q96CA5 | |
| BIRC7 | |
| Recombinant full length Human ML-IAP/BIRC7. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction