missing translation for 'onlineSavingsMsg'
Learn More

Livin Antibody, Alexa Fluor™ 700 , Novus Biologicals™

Product Code. 18433139 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18433139 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18433139 Supplier Novus Biologicals Supplier No. NBP297074AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Livin Polyclonal antibody specifically detects Livin in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Livin
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Livin (AAH14475.1; Met1-Ser298).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 79444
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.