missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Livin Antibody (88C570) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 22 publications
£177.00 - £422.00
Specifications
| Antigen | Livin |
|---|---|
| Clone | 88C570 |
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 2-5 ug/ml. Use reported in scientific literature (Nachmias et al (2007)) |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18067345
|
Novus Biologicals
NB100-56548SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18798773
|
Novus Biologicals
NB100-56548 |
0.1 mg |
£422.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Livin Monoclonal specifically detects Livin in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| Livin | |
| 0.5 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Zinc Finger | |
| Q96CA5 | |
| 79444 | |
| Recombinant human livin protein was used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 88C570 | |
| Western Blot 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 2-5 ug/ml. Use reported in scientific literature (Nachmias et al (2007)) | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 | |
| BIRC7 | |
| IgG1 κ | |
| Protein G purified | |
| The Livin antibody, clone 88C570 has been shown to recognize both Livin alpha (∼39 kDa) and Livin beta (∼37 kDa) isoforms (Nachmias et al, 2003). 2. The antibody also recognizes the 30 kDa (p30-Livin alpha) and 28 kDa (p28-Livin beta) apoptotic cleavage fragments of Livin alpha and beta, respectively (Nachmias et al, 2003). Apoptotic cleavage fragments of Livin have also been referred to as truncated livin (tLivin) (Nachmias et al, 2007). 3. The specificity of the antibody has been validated using Livin shRNA (Quiping et al, 2004). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title