missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Liver Perfusion Medium (1X)
GREENER_CHOICE

Product Code. 11540536
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Product Code. Quantity unitSize
11540536 500 mL 500mL
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11540536

Brand: Gibco™ 17701038

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Liver Perfusion Medium (1X)

Liver Perfusion Medium is a buffered, balanced salt solution formulated to cleanse the liver of blood, prevent clotting, and initiate loosening of cell-to-cell contact.
TRUSTED_SUSTAINABILITY

Specifications

Cell Type Hepatocyte Cells
Classification Serum-free
Form Liquid
Product Type Liver Perfusion Medium
Serum Level Serum-free
Without Additives No Glutamine, No Phenol Red
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.